Caprinaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: Decylaldehyde, Decanal, Decyl aldehyde

A953470-100G A953470-10G A953470-25G A953470-500G A953470-1000G
76698-822EA 19 USD
76698-822 76698-824 76698-826 76698-828 77627-778
Caprinaldehyde 97%
Caprinaldehyde

Caution: for Research Use Only

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 207
Melting Pt: 7
MDL Number: MFCD00007031
CAS Number: 112-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR