Bromothymol blue 98%

Supplier: Ambeed
Warning

Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3′,3″-Dibromothymolsulfonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

A921813-100G A921813-25G A921813-5G A921813-500G
76698-706EA 114 USD
76698-706 76698-708 77603-306 77627-760
Bromothymol blue 98%
Bromothymol blue

Caution: for Research Use Only

Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
MDL Number: MFCD00005872
CAS Number: 76-59-5
Merck Index: 13,01430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR