Bromocresol green sodium salt 98+%

Supplier: Ambeed
Warning

Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate

A841392-100G A841392-25G A841392-5G
76698-322EA 167 USD
76698-322 76698-324 76698-326
Bromocresol green sodium salt 98+%
Bromocresol green sodium salt

Caution: for Research Use Only

Formula: C₂₁H₁₃Br₄NaO₅S
MDL Number: MFCD00148898
CAS Number: 62625-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR