Bromothymol blue sodium salt Indicator Transition range: pH 5.8 - 7.6 yellow - blue

Supplier: Ambeed
Warning

Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulfonate, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate

A819808-100G A819808-25G A819808-5G
76698-158EA 52 USD
76698-158 76698-160 77878-674
Bromothymol blue sodium salt Indicator Transition range: pH 5.8 - 7.6 yellow - blue
Bromothymol blue sodium salt

Caution: for Research Use Only

Formula: C₂₇H₂₇Br₂NaO₅S
MW: 646.37 g/mol
MDL Number: MFCD00077263
CAS Number: 34722-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR