2',3'-Dideoxyguanosine 95%

Supplier: Ambeed
Warning

A783908-100MG A783908-250MG
76697-918EA 87.99 USD
76697-918 76697-920
2',3'-Dideoxyguanosine 95%
2',3'-Dideoxyguanosine

Caution: for Research Use Only

Formula: C₁₀H₁₃N₅O₃
MDL Number: MFCD00057074
CAS Number: 85326-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR