Benzyltriethylammonium iodide 98%

Supplier: Ambeed
Warning

A777107-25G A777107-5G A777107-1G
76697-862EA 81 USD
76697-862 76697-864 77878-622
Benzyltriethylammonium iodide 98%
Benzyltriethylammonium iodide

Caution: for Research Use Only

Formula: C₁₃H₂₂IN
MDL Number: MFCD00043178
CAS Number: 5400-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR