D-(-)-Isoascorbic acid 98%

Supplier: Ambeed
Warning

Synonyms: Erythorbic acid, (R)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, D-Isoascorbic acid

A775963-1000G A775963-100G A775963-25G A775963-500G
76697-848EA 70 USD
76697-848 76697-850 76697-852 77602-992
D-(-)-Isoascorbic acid 98%
D(-)-Isoascorbic acid

Caution: for Research Use Only

Formula: C₆H₈O₆
MDL Number: MFCD00005378
CAS Number: 89-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR