Copper (I) sulfide 99+%

Supplier: Ambeed
Warning

A752420-100G A752420-25G A752420-5G
76697-734EA 52 USD
76697-734 76697-736 76697-738
Copper (I) sulfide 99+%
Copper (I) sulphide

Caution: for Research Use Only

Formula: Cu₂S
MDL Number: MFCD00016062
CAS Number: 22205-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR