Choline chloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

A721479-25G A721479-500G A721479-5G A721479-1000G A721479-100G
77552-232EA 21.63 USD
77552-232 76697-504 76697-500 76697-502 76697-498
Choline chloride 97%
Choline chloride

Caution: for Research Use Only

Formula: HOH₂CCH₂N(CH₃)₃Cl
MW: 139.62 g/mol
MDL Number: MFCD00011721
CAS Number: 67-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR