Auramine O 80%

Supplier: Ambeed
Danger

Synonyms: Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow

A667033-25G A667033-1000G A667033-100G A667033-500G
76697-108EA 6 USD
76697-108 77627-334 77627-336 77627-338
Auramine O 80%
Auramine O

Caution: for Research Use Only

Formula: C₁₇H₂₂ClN₃
MW: 303.84 g/mol
MDL Number: MFCD00012484
CAS Number: 2465-27-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR