Apronal 99%

Supplier: Ambeed
Warning

Synonyms: N-Carbamoyl-2-isopropyl-4-pentenamide(2-Isopropyl-4-pentenoyl)ureaAllylisoprorylacetylureaApronalideAllylisopropylacetylcarbamideAllylisopropylacetylurea

A634600-100MG A634600-1MG A634600-1G
76696-856EA 5.31 USD
76696-856 76696-854 76696-852
Apronal 99%
Apronal

Caution: for Research Use Only

Formula: C₉H₁₆N₂O₂
MW: 184.24 g/mol
MDL Number: MFCD00210239
CAS Number: 528-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR