Methyl arachidate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl eicosanoate, Methyl Arachidate
A587624-100MG
A587624-1G
A587624-5G
A587624-25G
A587624-100G
A587624-250MG
76696-432EA
5
USD
76696-432
76696-434
76696-436
77551-766
77627-218
77878-387
Methyl arachidate 98%
Methyl arachidate
Caution: for Research Use Only
Formula:
C₂₁H₄₂O₂ MW: 326.56 g/mol |
MDL Number:
MFCD00009014 CAS Number: 1120-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...