Barium carbonate 99%

Supplier: Ambeed
Warning

A576014-1000G A576014-100G A576014-500G
76696-364EA 13 USD
76696-364 76696-366 76696-368
Barium carbonate 99%
Barium carbonate

Caution: for Research Use Only

Formula: BaCO₃
MW: 197.34 g/mol
Melting Pt: 1350
MDL Number: MFCD00003448
CAS Number: 513-77-9
Merck Index: 13,00972

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR