Bis(trimethylsilyl)acetylene 97%
Supplier: Ambeed
|
Danger
|
A565065-10G
A565065-1G
A565065-25G
A565065-5G
76696-248EA
17
USD
76696-248
76696-250
76696-252
76696-254
Bis(trimethylsilyl)acetylene 97%
Bis(trimethylsilyl)acetylene
Caution: for Research Use Only
Formula:
C₈H₁₈Si₂ |
MDL Number:
MFCD00008276 CAS Number: 14630-40-1 UN: 1993 ADR: 3,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...