Aminoacetaldehyde diethyl acetal 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal
A544100-10G
A544100-25G
A544100-5G
76696-062EA
6
USD
76696-062
76696-064
76696-066
Aminoacetaldehyde diethyl acetal 97%
Aminoacetaldehyde diethyl acetal
Caution: for Research Use Only
Formula:
C₆H₁₅NO₂ |
MDL Number:
MFCD00008136 CAS Number: 645-36-3 UN: 1989 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...