Allylchloro[1,3-bis(2,6-di-i-propylphenyl)-4,5-dihydroimidazol-2-ylidene]palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: Allyl(1,3-bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene)chloropalladate(II), Umicore CX23, (SIPr)Pd(allyl)Cl, Allyl[1,3-bis(2,6-diisopropylphenyl)-2-imidazolidinylidene]chloropalladium(II), Allylchloro-[1,3-bis-(2,6-diisopropylphenyl)-2-imidazolidinylidene]palladium(II), Allylchloro[1,3-bis-(2,6-diisopropylphenyl)imidazol-2-idinylidene]palladium(II), Allyl[1,3-bis(2,6-diisopropylphenyl)-2-imidazolidinylidene]chloropalladium(II)

A521838-100MG A521838-1G A521838-250MG A521838-5G
76695-848EA 18 USD
76695-848 76695-850 76695-852 77627-062
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)-4,5-dihydroimidazol-2-ylidene]palladium(II) 98%
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)-4,5-dihydroimidazol-2-ylidene]palladium(II)

Caution: for Research Use Only

Formula: C₃₀H₄₃ClN₂Pd
MDL Number: MFCD07782017
CAS Number: 478980-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR