Allyltris(trimethylsilyloxy)silane 97%

Supplier: Ambeed
Warning

Synonyms: 3-Allyl-1,1,1,5,5,5-hexamethyl-3-((trimethylsilyl)oxy)trisiloxane

A464633-1G A464633-250MG A464633-5G
76695-378EA 46 USD
76695-378 76695-380 76695-382
Allyltris(trimethylsilyloxy)silane 97%
Allyltris(trimethylsilyloxy)silane

Caution: for Research Use Only

Formula: C₁₂H₃₂O₃Si₄
MDL Number: MFCD00054804
CAS Number: 7087-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR