Calcium chloride 96%

Supplier: Ambeed
Warning

A460874-100G A460874-500G
76695-324EA 9.55 USD
76695-324 76695-326 76695-328
Calcium chloride 96%
Calcium chloride

Caution: for Research Use Only

Formula: CaCl₂
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR