Acetoxyacetyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: Acetoxyacetylchloride2-Chloro-2-oxoethyl acetate

A446762-25G A446762-1G
76695-190EA 5.15 USD
76695-190 76695-192
Acetoxyacetyl chloride 95%
Acetoxyacetyl chloride

Caution: for Research Use Only

Formula: C₄H₅ClO₃
MW: 136.53 g/mol
MDL Number: MFCD00011535
CAS Number: 13831-31-7
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR