Ammonium formate 98%

Supplier: Ambeed
Warning

Synonyms: Formic acid ammonium salt

A414767-100G A414767-25G A414767-500G A414767-1000G
76694-900EA 6 USD
76694-900 76694-902 76694-904 77551-240
Ammonium formate 98%
Ammonium formate

Caution: for Research Use Only

Formula: HCO₂NH₄
MW: 63.06 g/mol
MDL Number: MFCD00013103
CAS Number: 540-69-2
Merck Index: 13,00525

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR