Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Allyl[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]chloropalladium(II), (NHC)Pd(allyl)Cl, Allylchloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]palladium(II), CX 21
A396074-100MG
A396074-250MG
A396074-5G
A396074-1G
76694-732EA
21
USD
76694-732
76694-734
76694-736
77626-808
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II) 98%
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II)
Caution: for Research Use Only
Formula:
C₃₀H₄₁ClN₂Pd MW: 571.5330 g/mol |
MDL Number:
MFCD08561131 CAS Number: 478980-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...