Bis(4-fluorophenyl) sulfone 97%
Supplier: Ambeed
|
Warning
|
A383021-25G
A383021-500G
A383021-5G
A383021-100G
76694-506EA
19
USD
76694-506
76694-508
76694-510
77572-138
Bis(4-fluorophenyl) sulfone 97%
Bis(4-fluorophenyl) sulphone
Caution: for Research Use Only
Formula:
C₁₂H₈F₂O₂S |
MDL Number:
MFCD00000350 CAS Number: 383-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...