Acemetacin 98%

Supplier: Ambeed
Danger

Synonyms: [1-(4-Chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetoxyacetic Acid, 2-(2-(1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetoxy)acetic acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic Acid Carboxymethyl Ester

A374323-100MG A374323-1G
76694-376EA 5 USD
76694-376 76694-378
Acemetacin 98%
Acemetacin

Caution: for Research Use Only

Formula: C₂₁H₁₈ClNO₆
MDL Number: MFCD00151473
CAS Number: 53164-05-9
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR