α-Terpinene 95%

Supplier: Ambeed
Danger

Synonyms: 1-Isopropyl-4-methylcyclohexa-1,3-dienep-Mentha-1,3-diene

A370017-5G A370017-25G
76694-312EA 6.37 USD
76694-312 76694-310
α-Terpinene 95%
α-Terpinene

Caution: for Research Use Only

Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00001534
CAS Number: 99-86-5
UN: 2319
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR