3-Amino-4-methylbenzotrifluoride 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methyl-5-(trifluoromethyl)aniline

A361715-1G A361715-25G A361715-5G
76694-204EA 17 USD
76694-204 76694-206 76694-208
3-Amino-4-methylbenzotrifluoride 98%
3-Amino-4-methylbenzotrifluoride

Caution: for Research Use Only

Formula: C₈H₈F₃N
MW: 175.16 g/mol
MDL Number: MFCD00134580
CAS Number: 25449-96-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR