Erioglaucine disodium salt 85%
Supplier: Ambeed
|
Danger
|
Synonyms:
Sodium 2-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulfonate, FD and C Blue No. 1, Brilliant Blue FCF, Erioglaucin A, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonate, Alphazurine FG, Erioglaucine sodium salt, CI 42090, Acid blue 9, Food Blue No.1
A355767-10G
A355767-25G
76694-140EA
15
USD
76694-140
76694-142
Erioglaucine disodium salt 85%
Erioglaucine disodium salt
Caution: for Research Use Only
Formula:
C₃₇H₃₄Na₂N₂O₉S₃ MW: 792.84 g/mol |
MDL Number:
MFCD00012141 CAS Number: 3844-45-9 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...