Copper(II) ethylacetoacetate 98%

Supplier: Ambeed
Warning

A330781-1G A330781-25G A330781-5G
76693-886EA 17 USD
76693-886 76693-888 76693-890
Copper(II) ethylacetoacetate 98%
Copper(II) ethylacetoacetate

Caution: for Research Use Only

Formula: (CH₃CH₂CO₂CHCOCH₃)₂Cu
MDL Number: MFCD00017759
CAS Number: 14284-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR