Diammonium tetrachloropalladate Pd 36% min

Supplier: Ambeed
Warning

Synonyms: Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)

A309867-1G A309867-250MG A309867-5G
76693-544EA 48 USD
76693-544 76693-546 77626-546
Diammonium tetrachloropalladate Pd 36% min
Diammonium tetrachloropalladate

Caution: for Research Use Only

Formula: H₈Cl₄N₂Pd
MW: 284.31
Density: 2.17
MDL Number: MFCD00010884
CAS Number: 13820-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR