9-Aminoacridine hydrochloride monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: 9-Aminoacridine hydrochloride hydrate, Acridin-9-amine hydrochloride hydrate

A300216-1G A300216-25G A300216-5G
76693-392EA 9 USD
76693-392 76693-394 76693-396
9-Aminoacridine hydrochloride monohydrate 98%
9-Aminoacridine hydrochloride monohydrate

Caution: for Research Use Only

Formula: C₁₃H₁₁ClN₂·1H₂O
MDL Number: MFCD00150071
CAS Number: 52417-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR