Azelaoyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: Azelaoyl Chloride, Azelaoylchloride

A289732-5G A289732-25G
76693-222EA 26 USD
76693-222 77626-496
Azelaoyl chloride 95%
Azelaoyl chloride

Caution: for Research Use Only

Formula: C₉H₁₄Cl₂O₂
MW: 225.11 g/mol
MDL Number: MFCD00000767
CAS Number: 123-98-8
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR