Creatine (N-amidinosarcosine) monohydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate
A246444-100G
A246444-25G
A246444-500G
A246444-1000G
76692-372EA
16
USD
76692-372
76692-374
76692-376
77626-302
Creatine (N-amidinosarcosine) monohydrate 97%
Creatine (N-Amidinosarcosine) monohydrate
Caution: for Research Use Only
Formula:
C₄H₉N₃O₂·1H₂O MW: 149.15 g/mol (131.13 anhy) |
MDL Number:
MFCD00071582 CAS Number: 6020-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...