Piperidin-1-yl(quinoxalin-6-yl)methanone 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Piperidin-1-ylcarbonyl)quinoxaline, BDP 12, CX516, Ampalex, Ampakine CX516, 1-Piperidinyl-6-quinoxalinyl-methanone

A240364-100MG A240364-10MG A240364-1MG A240364-250MG A240364-50MG A240364-5MG
76692-260EA 86 USD
76692-260 76692-262 76692-264 76692-266 76692-268 76692-270
Piperidin-1-yl(quinoxalin-6-yl)methanone 97%
Piperidin-1-yl(quinoxalin-6-yl)methanone

Caution: for Research Use Only

Formula: C₁₄H₁₅N₃O
MDL Number: MFCD00943201
CAS Number: 154235-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR