9-Bromo-10-(2-naphthyl)anthracene 97%

Supplier: Ambeed
Warning

A238257-1G A238257-25G A238257-5G
76692-216EA 10 USD
76692-216 76692-218 76692-220
9-Bromo-10-(2-naphthyl)anthracene 97%
9-Bromo-10-(2-naphthyl)anthracene

Caution: for Research Use Only

Formula: C₂₄H₁₅Br
MDL Number: MFCD09832882
CAS Number: 474688-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR