Auraptene 99%

Supplier: Ambeed
Warning

Synonyms: (E)-7-((3,7-Dimethylocta-2,6-dien-1-yl)oxy)-2H-chromen-2-one, 7-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-2H-chromen-2-one, 7-Geranyloxycoumarin

A235149-1MG A235149-250MG A235149-50MG
76692-140EA 7 USD
76692-140 76692-142 76692-144
Auraptene 99%
Auraptene

Caution: for Research Use Only

Formula: C₁₉H₂₂O₃
MW: 298.4 g/mol
MDL Number: MFCD00075948
CAS Number: 495-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR