8-Bromo-2'-deoxyguanosine 98%

Supplier: Ambeed
Warning

A217092-100MG A217092-1G A217092-250MG A217092-5G A217092-25G
76691-664EA 8 USD
76691-664 76691-666 76691-668 76691-670 77626-100
8-Bromo-2'-deoxyguanosine 98%
8-Bromo-2'-deoxyguanosine

Caution: for Research Use Only

Formula: C₁₀H₁₂BrN₅O₄
MDL Number: MFCD01630971
CAS Number: 13389-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR