5-Chloro-N-ethyl-4-hydroxy-1-methyl-2-oxo-N-phenyl-1,2-dihydroquinoline-3-carboxamide 98%

Supplier: Ambeed
Warning

A216404-100MG A216404-10MG A216404-1G A216404-1MG A216404-250MG A216404-2MG A216404-50MG A216404-5G A216404-5MG
76691-630EA 48 USD
76691-630 76691-632 76691-634 76691-636 76691-638 76691-640 76691-642 76691-644 76691-646
5-Chloro-N-ethyl-4-hydroxy-1-methyl-2-oxo-N-phenyl-1,2-dihydroquinoline-3-carboxamide 98%
5-Chloro-N-ethyl-4-hydroxy-1-methyl-2-oxo-N-phenyl-1,2-dihydroquinoline-3-carboxamide

Caution: for Research Use Only

Formula: C₁₉H₁₇ClN₂O₃
MDL Number: MFCD08689859
CAS Number: 248281-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR