Chlorocyclohexane 98%

Supplier: Ambeed
Danger

Synonyms: Cyclohexyl chloride

A211405-25G A211405-100G
76691-506EA 14 USD
76691-506 77593-386
Chlorocyclohexane 98%
Chlorocyclohexane

Caution: for Research Use Only

Formula: C₆H₁₁Cl
MDL Number: MFCD00003822
CAS Number: 542-18-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR