Aliskiren hemifumarate 98%

Supplier: Ambeed
Warning

Synonyms: Rasilez, (2S,4S,5S,7S)-N-(2-Carbamoyl-2-methylpropyl)-5-amino-4-hydroxy-2,7diisopropyl-8-[4-methoxy-3-(3-methoxypropoxy)phenyl]-octanamide hemifumarate, Aliskiren Hemifumarate, SPP-100, CGP-60536, (2S,4S,5S,7S)-N-(2-Carbamoyl-2-methylpropyl)-5-amino-4-hydroxy-2,7diisopropyl-8-[4-methoxy-3-(3-methoxypropoxy)phenyl]-octanamide hemifumarate, CGP60536B, (2S,4S,5S,7S)-5-Amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-4-hydroxy-2-isopropyl-7-(4-methoxy-3-(3-methoxypropoxy)benzyl)-8-methylnonanamide hemifumarate

A201324-100MG A201324-10MG A201324-1MG A201324-50MG A201324-1G A201324-250MG
76691-188EA 64 USD
76691-188 76691-190 76691-192 76691-194 77626-006 77626-008
Aliskiren hemifumarate 98%
Aliskiren hemifumarate

Caution: for Research Use Only

Formula: C₃₀H₅₃N₃O₆·0.5C₄H₄O₄
MDL Number: MFCD27756642
CAS Number: 173334-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR