Aluminium distearate 99% (4.5-6% Al basis)

Supplier: Ambeed
Warning

A200337-100G A200337-500G A200337-25G
76691-156EA 9 USD
76691-156 76691-158 77877-588
Aluminium distearate 99% (4.5-6% Al basis)
Aluminium distearate

Caution: for Research Use Only

Formula: C₃₆H₇₁AlO₅
MDL Number: MFCD00055007
CAS Number: 300-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR