Cromolyn sodium 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Disodium Cromoglycate
, Sodium 5,5'-((2-hydroxypropane-1,3-diyl)bis(oxy))bis(4-oxo-4H-chromene-2-carboxylate)
, 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis(4-oxo-4H-1-benzopyran-2-carboxylic Acid) Disodium Salt
A194389-1G
A194389-25G
A194389-50MG
A194389-10MG
A194389-5G
A194389-10G
76691-008EA
6.18
USD
76691-008
76691-006
76691-004
76691-002
76691-000
76691-010
Cromolyn sodium 97%
Cromolyn sodium
Caution: for Research Use Only
Formula:
C₂₃H₁₆O₁₁ MW: 512.33 g/mol |
MDL Number:
MFCD22575281 CAS Number: 15826-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...