Alibendol 98%

Supplier: Ambeed
Warning

Synonyms: Cebera, 5-Allyl-2-hydroxy-N-(2-hydroxyethyl)-3-methoxybenzamide, 5-Allyl-N-(2-hydroxyethyl)-3-methoxysalicylamide

A192206-100G A192206-250MG A192206-25G A192206-5G
76690-928EA 114 USD
76690-928 76690-930 76690-932 76690-934
Alibendol 98%
Alibendol

Caution: for Research Use Only

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
MDL Number: MFCD00865165
CAS Number: 26750-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR