Dehydroacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

A177106-500G A177106-10G A177106-100G A177106-1000G A177106-25G
76690-396EA 9.55 USD
76690-396 76690-398 76690-400 76690-402 76690-394
Dehydroacetic acid 98%
Dehydracetic acid

Caution: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00066709
CAS Number: 520-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR