(S)-(+)-O-Acetylmandelic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-Acetoxy-2-phenylacetic acid, (S)-(+)-α-Acetoxyphenylacetic Acid, (+)-O-Acetyl-L-mandelic Acid
A145410-100G
A145410-1G
A145410-500G
A145410-5G
A145410-25G
76689-136EA
21
USD
76689-136
76689-138
76689-140
76689-142
76689-144
77299-452
(S)-(+)-O-Acetylmandelic acid 97%
(S)-(+)-O-Acetylmandelic acid
Caution: for Research Use Only
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol |
MDL Number:
MFCD00064215 CAS Number: 7322-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...