6-O-Palmitoyl-L-ascorbic acid 95%
Supplier: Ambeed
|
Warning
|
A142721-100G
A142721-10G
A142721-250MG
A142721-25G
A142721-500G
76689-050EA
35
USD
76689-050
76689-052
76689-054
76689-056
76689-058
6-O-Palmitoyl-L-ascorbic acid 95%
6-O-Palmitoyl-L-ascorbic acid
Caution: for Research Use Only
Formula:
C₂₂H₃₈O₇ |
MDL Number:
MFCD00005377 CAS Number: 137-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...