Cyclohexylméthanol 98%

Supplier: Ambeed
Warning

A140963-100G A140963-10G A140963-500G A140963-25G
76689-010EA 18.04 USD
76689-010 76689-012 76689-014 77316-050
Cyclohexylméthanol 98%
Cyclohexylméthanol

Caution: for Research Use Only

Formula: C₇H₁₄O
MDL Number: MFCD00001510
CAS Number: 100-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR