Ambrisentan 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic acid, (S)-2-((4,6-Dimethylpyrimidin-2-yl)oxy)-3-methoxy-3,3-diphenylpropanoic acid

A121424-100MG A121424-10MG A121424-1G A121424-250MG A121424-50MG A121424-5G A121424-5MG
76688-122EA 27 USD
76688-122 76688-124 76688-126 76688-128 76688-130 76688-132 77599-748
Ambrisentan 98%
Ambrisentan

Caution: for Research Use Only

Formula: C₂₂H₂₂N₂O₄
MW: 378.42 g/mol
MDL Number: MFCD09842330
CAS Number: 177036-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR