Allyl cinnamate 98% stabilised

Supplier: Ambeed
Warning

Synonyms: Allyl Cinnamate, Cinnamic Acid Allyl Ester, Cinnamic acid Allyl Ester

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A118025-100G A118025-10G A118025-25G
76687-982EA 17 USD
76687-982 76687-984 77876-864
Allyl cinnamate 98% stabilised
Allyl cinnamate

Caution: for Research Use Only

Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
MDL Number: MFCD00026105
CAS Number: 1866-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR