7-Chloroquinoline 97%

Supplier: Ambeed
Warning

A105655-100MG A105655-1G A105655-250MG A105655-25G
76687-236EA 5 USD
76687-236 76687-238 76687-240 77876-808
7-Chloroquinoline 97%
7-Chloroquinoline

Caution: for Research Use Only

Formula: C₉H₆ClN
MDL Number: MFCD00956370
CAS Number: 612-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR