Anti-CD4 Rabbit Monoclonal Antibody [clone: 766]

Supplier: Sino Biological
50134-R766-100 50134-R766-20
76644-068EA 269 USD
76644-068 76777-434
Anti-CD4 Rabbit Monoclonal Antibody [clone: 766]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD4 (rM CD4; Catalog No: 50134-M08H; NP_038516.1; Met1-Thr394).

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 766
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR