Anti-CXCL8 Mouse Monoclonal Antibody [clone: 5]

Supplier: Sino Biological
10098-MM05-100 10098-MM05-20
76644-042EA 299 USD
76644-042 76778-662
Anti-CXCL8 Mouse Monoclonal Antibody [clone: 5]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-8/Interleukin-8/CXCL8 (rh IL-8/Interleukin-8/CXCL8; Catalog No: 10098-H08Y; NP_000575.1; Ser28-Ser99). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR